missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin/Proinsulin Antibody (D3E7cc) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB200-435JF669
3060.00 NOK valid until 2025-12-16
Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Insulin/Proinsulin Monoclonal antibody specifically detects Insulin/Proinsulin in Bovine,Human,Porcine,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Frozen),Sandwich ELISA
Specifications
| Insulin/Proinsulin | |
| Monoclonal | |
| Janelia Fluor 669 | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with native human insulin. | |
| 0.2 mL | |
| Diabetes Research | |
| 3630 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Sandwich ELISA | |
| D3E7cc | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Bovine, Human, Pig, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction