Learn More
Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing
Product Code. 10234231
Click to view available options
Range:
0 to 500 mg/L Total
0.00 to 10.00 mg/L Free and Total
0.00 to 5.00 mg/L Total
0.000 to 0.500 mg/L Free
No. of Tests:
300
600
For Use With (Application):
Chlorine Ultra High Range
Free Chlorine Ultra Low Range
Free and Total Chlorine High Range
Total Chlorine
Total Chlorine (DPD3)
Unit Size:
Each
Product Code. 10234231
Brand: Hanna Instruments™ HI9371103
Only null left
Add to basket
Add to basket
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Chlorine Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
These high quality reagents are designed to follow accepted test methods for colorimetric determinations of total and/or free chlorine. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
  • Reagent sets available for: Free and total chlorine (high range), Free chlorine (ultra low range), Total chlorine (ultra high range, ultra low range, undetermined range)
Specifications
For Use With (Application) Total Chlorine
For Use With (Equipment) Compatible Hanna benchtop and portable photometers
Includes Individual Packets
No. of Tests 300
Quantity 1 Set
Range 0.00 to 5.00 mg/L Total
Test Method USEPA method 330.5 and Standard Method 4500-Cl G
Type Water Testing Kit
compliance-icons compliance-icons
Product Identifier
  • Total Chlorine Reagents (300 tests)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 1
Hazard Statement
  • H332-Harmful if inhaled.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : potassium iodide, EDTA