Learn More
Hanna Instruments™ Potassium Reagent Sets, For Colorimetric Water Testing
Product Code. 11672660
Click to view available options
Quantity:
1 Set
Unit Size:
30mL
Product Code. 11672660
Brand: Hanna Instruments™ HI9375001
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Potassium Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
These high quality reagents are designed to follow an adaptation of the Tetraphenylborate method for colorimetric determination of potassium. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
  • Pre-made, pre-measured water testing reagents for ease of use
  • Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
  • Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
Analytes Tetraphenylborate
For Use With (Application) Potassium determination
For Use With (Equipment) Compatible Hanna benchtop and portable photometers
Includes Dropper bottles and individual packets
No. of Tests 100
Quantity 1 Set
Range 0.0 to 200mg/L potassium (benchtop photometers) and 0.0 to 100mg/L potassium (portable photometer)
Test Method Cholorimetric determination
Type Water Testing Kit
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • POTASSIUM REAGENT FOR 16428
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H350-May cause cancer.
  • H332-Harmful if inhaled.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
  • H318-Causes serious eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/first aider.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/first aider.
  • P370+P378-In case of fire: Use to extinguish.
Supplemental information
  • MIXTURE LIST-Contains : edta disodium salt,formaldehyde, methat, sodium tetraphenylborate/ formaldehyde, methanol