Learn More
Hamilton™ Terasaki Dispenser
Description
- Designed for rapid filling of 60 well HLA/Terasaki plates with 6.35mm well spacing
- Uses removable needles
- Available for a range of volumes
Specifications
Specifications
| Quantity | 1/Ea. |
| Type | Dispenser |
| Material | Borosilicate Glass, PTFE |
| Capacity (Metric) | 250 μL |