Learn More
Instrument Check Standard 10 For Environmental Analysis, SPEX CertiPrep™
Product Code. 11971626 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11971626 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11971626 Supplier SPEX Certiprep Supplier No. CALMIX10500
  • ONLINE EXCLUSIVE PRICE
    4535.00 NOK
    4303.37 NOK / 500mL
    Save 231.63 NOK
    5% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
AnalyteConcentration
K500μg/mL
Al50μg/mL
Ba50μg/mL
Cd50μg/mL
Cu50μg/mL
Mn50μg/mL
Zn50μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging Triple-leached LDPE bottle
CAS 7446-07-3
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Molecular Formula O2Te
MDL Number MFCD00011263
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
InChI Key LAJZODKXOMJMPK-UHFFFAOYSA-N
Type Certified Reference Material
SMILES O=[Te]=O
IUPAC Name tellurium dioxide
Solvent or Matrix 2% HNO3
Molecular Weight (g/mol) 159.60
PubChem CID 62638
Quantity 500 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Components in 2% HNO3:
Al    •50Glc mug/mL
Mn    •50Glc mug/mL
Ba    •50Glc mug/mL
K    •500Glc mug/mL
Cd    •50Glc mug/mL
Zn    •50Glc mug/mL
Cu    •50Glc mug/mL
Chemical Name or Material Instrument Check Standard 10
Show More Show Less
compliance-icons
Product Identifier
  • Instrument Check Standard 10
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid
Product intended for laboratory use only