Learn More
3M™ Instant Microbe Removing Wipes
Description
- Quickly traps, holds and removes 99.9% of microbes, including SARS-CoV-2*
- Virtually eliminates cross contamination
- Works using only water – no disinfectant or specific PPE required
- Easily removes as you wipe, no dwell time
- Also works with quat disinfectants
- Refillable containers to reduce waste
- If wipes dry out, simply add water
Specifications
Specifications
| Product Type | Wipe |
| Length (English) | 7.7 in. |
| Length (Metric) | 195 mm |
| Width (English) | 10 in. |
| Width (Metric) | 254 mm |
| Certifications/Compliance | FR/GE |
| Dimensions (L x W) | 195 mm x 254 mm |
| For Use With (Application) | Microbe Removal |
| Packaging Quantity | 85 Sheets/Canister, 4 Canisters/Case |
*Based on third party testing with water only.