missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSRR/IR-related receptor Mouse anti-Human, Alexa Fluor 700, Clone: 326929, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB3055N-100UG
This item is not returnable.
View return policy
Description
INSRR/IR-related receptor Monoclonal specifically detects INSRR/IR-related receptor in Human samples. It is validated for Flow Cytometry.Specifications
| INSRR/IR-related receptor | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P14616 | |
| INSRR | |
| Mouse myeloma cell line NS0-derived recombinant human INSRR Leu24-Leu923 Accession # P14616 | |
| RUO | |
| 3645 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 326929 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| EC 2.7.10, EC 2.7.10.1, insulin receptor-related protein, insulin receptor-related receptor, IRR, IRRIR-related receptor | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human INSRR in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only (RUO)