Learn More
Abeba™ Insoles 3565
Product Code. p-7147581
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
Unit Size:
1 pair
Product Code. Size unitSize
15640198 36 1 pair
15650198 37 1 pair
15660198 38 1 pair
15670198 39 1 pair
15680198 40 1 pair
15690198 41 1 pair
15600208 42 1 pair
7 options
Product Code. 15640198
Brand: Abeba™ 3565/36
Only null left
Add to basket
Add to basket
Massage knobs stimulate circulation. ESD dissipative and antistatic.
  • Microfiber
  • Anthracite
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Stimulating circulation by massage knobs, comfortable orthopaedic footbed
  • Reflexor massage active-footbed
Specifications
Color Anthracite
Material Microfiber
Size 36
Style 3565