Learn More
Ohaus™ Insert Retainer
Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
Vortexer
Unit Size:
Each
Description
- OHAUS offers a large selection of accessories that can be used on all OHAUS vortex mixers
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
Specifications
| Product Type | Insert Retainer |
| Material | Aluminum |
| Description | Insert Retainer |
| For Use With (Equipment) | Vortexer |