Learn More
Pyrex™ Tubing Insert for GL14 Screw Cap
Product Code. 15153937
Click to view available options
For Use With:
GL14 screwcap with tubing outlet, 1.6mm ID
GL14 screwcap with tubing outlet, 3.0mm ID
GL14 screwcap with tubing outlet, 3.2mm ID
GL14 screwcap with tubing outlet, 6.0mm ID
Unit Size:
Each
Product Code. 15153937
Brand: Pyrex™ 4539/06
Only null left
Add to basket
Add to basket
Choose a two or three port connection system screwcap and then add tubing adapters and inserts to attach tubing. Pyrex™ Tubing Insert for GL14 Screw Cap provides a means to transfer liquids within a closed, sterile system.
  • For use with Pyrex™ GL45 media-lab bottles
  • Facilitates the transfer of liquids within a closed sterile system
  • Autoclavable and dishwasher safe
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
For Use With GL14 screwcap with tubing outlet, 3.2mm ID
Max. Operating Temperature (Metric) 140°C
Product Type Tubing Insert