Learn More
Thermo Scientific Chemicals Inositol hexaphosphoric acid, 50 wt% aqueous solution
Product Code. p-148591 Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mL
1 L
2.5 L
missing translation for 'unitSize'
1 L
2.5 L
250mL
Product Code. 10562251
missing translation for 'mfr': Thermo Scientific Chemicals 235372500
Request Bulk or Custom Format
CAS: 83-86-3 | C6H18O24P6 | 660.03 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Inositol hexaphosphoric acid
Name Note 40-50 wt. % Aqueous Solution
CAS 83-86-3, 7732-18-5
Boiling Point 105.0°C
Molecular Formula C6H18O24P6
MDL Number MFCD00082309
Density 1.3700g/mL
Synonym phytic acid, phytate, fytic acid, inositol hexaphosphate, alkalovert, myo-inositol hexaphosphate, alkovert, phytine, acide fytique, acidum fyticum
Solubility Information Solubility in water: miscible.
InChI Key IMQLKJBTEOYOSI-GPIVLXJGSA-N
SMILES OP(O)(=O)O[C@H]1[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H]1OP(O)(O)=O
IUPAC Name (2,3,4,5,6-pentaphosphonooxycyclohexyl) dihydrogen phosphate
Molecular Weight (g/mol) 660.03
PubChem CID 890
Formula Weight 660.03
Packaging Glass bottle
Refractive Index 1.391
Specific Gravity 1.37
Quantity 250 mL
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Inositol hexaphosphoric acid, 40-50 wt% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.