Description
The iNOS antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human iNOS. This antibody recognizes human, mouse antigen. The iNOS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | iNOS |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 533 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 4843 |
| Target Species | Human, Mouse |
| Show More |