Learn More
BRAND™ Inoculation Loop with Needle
Product Code. 10272431 Shop All Brand Products
Change view
Click to view available options
Volume:
1 μL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Unit Size
10272431 1 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10272431 Supplier BRAND™ Supplier No. 452215
  • ONLINE EXCLUSIVE PRICE
    990.00 NOK
    682.40 NOK / Pack of 1000
    Save 307.60 NOK
    31% Off
Estimated Shipment: 18-12-2026
Inoculation Loop with Needle designed for inoculation of nutrient media.
  • For inoculation of nutrient media
  • Available with loop at one end and needle at the other
  • High flexibility permits gentle streaking without damaging the nutrient surface
  • γ-ray sterilized
TRUSTED_SUSTAINABILITY
Specifications
Volume 1 μL
Material Polystyrene
Sterility Gamma Sterilized
Color Yellow
Includes Needle
For Use With (Application) For inoculation of nutrient media