Learn More
New Brunswick Scientific™ Innova™ 42 Incubator Shakers
Product Code. 11350025
Change view
Click to view available options
Orbit:
19 mm
25 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit unitSize
11350025 25 mm Each
11330025 19 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 11350025 Supplier New Brunswick Scientific™ Supplier No. M13350012
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Use these versatile incubator shakers for a variety of applications. New Brunswick Scientific™ Innova™ 42 Incubator Shakers provide exceptional convenience allowing you to simultaneously shake and static incubate cultures under the same conditions.
  • Stackable or benchtop incubator shaker with optional refrigeration
  • Use on or under the bench, or double stacked to save space
  • Optional culture drawer, gassing manifold, remote alarm, and humidity monitor are available
  • Cast iron triple-eccentric counterbalanced drive provides years of dependable operation
  • Programmable Innova controls automatically changes temperature, speed and optional photosynthetic and UV germicidal lights at timed intervals
  • Large viewing window and internal light provide easy viewing of samples without the need to open the door
  • Built-in water reservoir humidifies chamber to reduce sample evaporation, while also protecting unit from spills and including a drain for easy cleaning
  • Versatile accessory platforms (sold separately) accommodates flasks up to 4 L as well as test tube racks and microplate holders
TRUSTED_SUSTAINABILITY
Specifications
Orbit 25 mm
Speed Range 25 to 400 rpm
Electrical Requirements 230 V, 50 Hz
Voltage 230 V
Holds 4 L
Product Type Incubator Shaker
Frequency 50 Hz
Depth (Metric) 75.4 cm
Height (Metric) 131 cm
Width (Metric) 62 cm
Model Innova 42
Timer Range 0.01 to 99:59 hr.