Learn More
3M™ Inner Protection Plates
Product Code. 11735304
Change view
Click to view available options
For Use With (Equipment):
Can be used to increase one dark shade of welding filter
Can be used to increase two dark shades of welding filter
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11735304 Can be used to increase two dark shades of welding filter Pack of 5
11745294 Can be used to increase one dark shade of welding filter Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11735304 Supplier 3M™ Supplier No. 7100080228
  • ONLINE EXCLUSIVE PRICE
    679.00 NOK
    516.90 NOK / Pack of 5
    Save 162.10 NOK
    23% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Specifications
Color Clear
Dimensions (L x W) 42 x 91 mm
For Use With (Equipment) Can be used to increase two dark shades of welding filter
For Use With (Application) Can be used to Increase two dark shades of welding filter.
Product Type Inner Protection Plate