Learn More
IKA™ Inlet Tube Condenser
Product Code. 11845912 Shop All IKA Products
Click to view available options
Type:
Diagonal Condenser
Vertical Condenser
Unit Size:
Each
Product Code. 11845912
Brand: IKA™ undefined
Only null left
Specifications
Material Glass, PTFE Inlet Tube
Type Vertical Condenser
Includes 1000mL Evaporator, 1000 ml receiving flask
Quantity 1 Each
Shape Vertical
Weight (Metric) 100 g