Learn More
Brady™ Inkjet Polyester Roll Labels with 76.2 mm Core
Product Code. p-7224579
Click to view available options
Width (Metric):
57.2 mm
102 mm
Unit Size:
Each
Product Code. 17595136
Brand: Brady™ 153647
Only null left
Add to basket
Add to basket
Inkjet-printable polyester tape featuring a specially-formulated topcoat for improved durability and color.
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol, and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Designed for roll inkjet printers
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Continuous
Material Polyester (B-2569)
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Electrical Labeling, Facility and General Identification, Hazard Identification, Warehouse Marking
For Use With (Equipment) J2000 and J4000 Inkjet Printers
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 1 Tape/Bag
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.1 mm
Dimensions (LxW) 15.24 m x 57.15 mm
Length (English) 49 ft.
Length (Metric) 15 m
Width (English) 2.25 in.
Width (Metric) 57.2 mm
Alternate Part Number JPC-2250-2569