Learn More
B Braun™ Injekt™-F Fine Dosage Syringe
Product Code. 10303002
Change view
Click to view available options
Volume (Metric):
1 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10303002 1 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10303002 Supplier B Braun™ Supplier No. 9166017V
  • ONLINE EXCLUSIVE PRICE
    333.08 NOK
    136.12 NOK / Pack of 100
    Save 196.96 NOK
    59% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Manufactured from polypropylene/polyethylene. B. Braun™ Injekt™-F Fine Dosage Syringe has high contrast black graduation for ideal readability.
    • Made of polypropylene/polyethylene
    • Green plunger
    • Highly transparent barrel
    • High contrast black graduation, permanent marking, ideal readability
    • Displacement spike: Reduces residual volume and minimizes unnecessary loss of medicine
    • Centric Luer cone
    • Safe plunger backstop
    • Easy aspiration up to maximum volume
    • Latex-free/PVC-free
    • Silicone oil-free
    • Single sterile-packed (in box of 100 pcs)
Specifications
Certifications/Compliance ISO 7886-1
Material Polyethylene, Polypropylene
Color Green Plunger
Description Black graduation
For Use With (Application) Ideal for simple injections, vaccinations and more
Latex Free Yes
Product Type Centric Luer Cone 2-part Syringe
Disposable Single-use
Quantity 100/Pk.
Volume (Metric) 1 mL
Connection Type Centric Luer Cone
Graduated Yes