Learn More
- Home
- Products
- Chromatography Supplies
- Chromatography Port Injection Liners
- Thermo Scientific™ LinerGOLD™ GC Liners
Thermo Scientific™ LinerGOLD™ GC Liners
Product Code. 10300875 Shop All Thermo Scientific Products| Product Code. | Diameter (Metric) Inner | Length (Metric) | Quantity | Product Type | Unit Size |
|---|---|---|---|---|---|
| 10300875 | 2 mm | 120 mm | 5 | PTV Baffle Liner (Siltek) | Pack of 5 |
| 10299954 | 2 mm | 120 mm | 2 | PTV Siltek Metal Liner | Pack of 2 |
| 10428854 | 1 mm | 120 mm | 2 | PTV Siltek Metal Liner | Pack of 2 |
| 11827310 | 1 mm | 120 mm | 2 | PTV Silcosteel Liner for OC | Pack of 2 |
| 10651324 | 5 mm | 105 mm | 5 | Split Straight Liner | Pack of 5 |
| 10572075 | 3 mm | 105 mm | 5 | Split Straight Liner | Pack of 5 |
| 10455173 | 5 mm | 105 mm | 5 | Splitless Straight Liner | Pack of 5 |
| 11827290 | 1 mm | 120 mm | 5 | PTV Straight Liner | Pack of 5 |
| 10651914 | 2 mm | 120 mm | 5 | PTV Straight Liner | Pack of 5 |
| 10209964 | 2 mm | 120 mm | 5 | PTV Liner, Sintered Lining | Pack of 5 |
| 10714847 | 1 mm | 120 mm | 5 | PTV Liner, Three Baffles | Pack of 5 |
| 11877350 | 0.75 mm | 105 mm | 5 | PTV Split/Splitless Straight Liner | Pack of 5 |
| 10732527 | 5 mm | 105 mm | 25 | Split Straight Liner | Pack of 25 |
| 10732147 | 3 mm | 105 mm | 25 | Split Straight Liner | Pack of 25 |
| 11520964 | 5 mm | 105 mm | 25 | Splitless Straight Liner | Pack of 25 |
| 10619865 | 1 mm | 120 mm | 25 | PTV Straight Liner | Pack of 25 |
| 10690616 | 2 mm | 120 mm | 25 | PTV Straight Liner | Pack of 25 |
| 10147694 | 1 mm | 120 mm | 25 | PTV Liner, Three Baffles | Pack of 25 |
| 10416113 | 5 mm | 105 mm | 5 | Split FocusLiner | Pack of 5 |
| 10594465 | 3 mm | 105 mm | 5 | Splitless Straight Liner (Siltek) | Pack of 5 |
| 10314273 | 5 mm | 105 mm | 5 | Splitless FocusLiner | Pack of 5 |
| 10751957 | 5 mm | 105 mm | 5 | Splitless FocusLiner for 50 mm Needle | Pack of 5 |
| 10489614 | 5 mm | 105 mm | - | Splitless FocusLiner | Pack of 25 |
| 10764247 | 5 mm | 105 mm | 25 | Split FocusLiner | Pack of 25 |
| 10761957 | 5 mm | 105 mm | 25 | Splitless FocusLiner | Pack of 25 |
| 10053004 | 2 mm | 120 mm | 5 | PTV Glass Straight Liner (Deactivated) | Pack of 2 |
| 10209174 | 2 mm | 120 mm | 2 | PTV Siltek Metal Liner, Wool | Pack of 2 |
| 15237498 | 4 mm | 78.5 mm | 5 | Split/Splitless Mini-Lam Liner | Pack of 5 |
| 15247498 | Mixed | Mixed | 5 | Split/Splitless Mixed Liner Sample Pack | Pack of 5 |
| 10190553 | 3 mm | 105 mm | 5 | Splitless Straight Liner | Pack of 5 |
| 15458000 | 5 mm | 105 mm | 1 | Direct Straight Liner | Each |
| 15488000 | 3 mm | 105 mm | 1 | Split Straight Liner | Each |
| 15418010 | 3 mm | 105 mm | 1 | Splitless Liner | Each |
| 15448010 | 5 mm | 105 mm | 1 | Splitless Liner | Each |
| 15468000 | 5 mm | 105 mm | 5 | Direct Straight Liner | Pack of 5 |
| 15498000 | 3 mm | 105 mm | 5 | Split Straight Liner | Pack of 5 |
| 15428010 | 3 mm | 105 mm | 5 | Splitless Liner | Pack of 5 |
| 15408000 | 4 mm | 78.5 mm | 5 | Precision Liner | Pack of 5 |
| 15438000 | 1 mm | 78.5 mm | 5 | Direct Straight Liner | Pack of 5 |
| 15457980 | 4 mm | 78.5 mm | 5 | Split/Splitless Liner | Pack of 5 |
| 15487980 | 4 mm | 78.5 mm | 5 | Split/Splitless Straight Liner | Pack of 5 |
| 15417990 | 4 mm | 78.5 mm | 5 | Splitless Liner, Single Taper | Pack of 5 |
| 15447990 | 4 mm | 78.5 mm | 5 | Splitless Liner, Single Taper, Wool | Pack of 5 |
| 15477990 | 4 mm | 78.5 mm | 5 | Splitless Liner, Double Taper | Pack of 5 |
| 15458010 | 5 mm | 105 mm | 5 | Splitless Liner | Pack of 5 |
| 15488010 | 5 mm | 105 mm | 5 | Precision Liner | Pack of 5 |
| 15478000 | 5 mm | 105 mm | 25 | Direct Straight Liner | Pack of 25 |
| 15408010 | 3 mm | 105 mm | 25 | Split Straight Liner | Pack of 25 |
| 15438010 | 3 mm | 105 mm | 25 | Splitless Liner | Pack of 25 |
| 15468010 | 5 mm | 105 mm | 25 | Splitless Liner | Pack of 25 |
| 15447980 | 4 mm | 78.5 mm | 1 | Split/Splitless Liner | Each |
| 15497990 | 4 mm | 78.5 mm | 1 | Precision Liner | Each |
| 15477980 | 4 mm | 78.5 mm | 1 | Split/Splitless Straight Liner | Each |
| 15428000 | 1 mm | 78.5 mm | 1 | Direct Straight Liner | Each |
| 15407990 | 4 mm | 78.5 mm | 1 | Splitless Liner, Single Taper | Each |
| 15467990 | 4 mm | 78.5 mm | 1 | Splitless Liner, Double Taper | Each |
| 15437990 | 4 mm | 78.5 mm | 1 | Splitless Liner, Single Taper | Each |
| 15418000 | 4 mm | 78.5 mm | 25 | Precision Liner | Pack of 25 |
| 15467980 | 4 mm | 78.5 mm | 25 | Split/Splitless Liner | Pack of 25 |
| 15497980 | 4 mm | 78.5 mm | 25 | Split/Splitless Straight Liner | Pack of 25 |
| 15448000 | 1 mm | 78.5 mm | 25 | Direct Straight Liner | Pack of 25 |
| 15427990 | 4 mm | 78.5 mm | 25 | Splitless Liner, Single Taper | Pack of 25 |
| 15487990 | 4 mm | 78.5 mm | 25 | Splitless Liner, Double Taper | Pack of 25 |
| 15457990 | 4 mm | 78.5 mm | 25 | Splitless Liner, Single Taper, Wool | Pack of 25 |
| 15438020 | 2 mm | 120 mm | 1 | PTV Liner, Concentric Baffles | Each |
| 15478010 | 5 mm | 105 mm | 1 | Precision Liner | Each |
| 15408020 | 5 mm | 105 mm | 1 | Splitless Precision Liner | Each |
| 15640249 | 1 mm | 120 mm | 5 | PTV Liner, Offset 5 Baffles | Pack of 5 |
| 15448020 | 2 mm | 120 mm | 5 | PTV Liner, Concentric Baffles | Pack of 5 |
| 15418020 | 5 mm | 105 mm | 5 | Splitless Precision Liner | Pack of 5 |
| 15458020 | 2 mm | 120 mm | 25 | PTV Liner, Concentric Baffles | Pack of 25 |
| 15498010 | 5 mm | 105 mm | 25 | Precision Liner | Pack of 25 |
| 15428020 | 5 mm | 105 mm | 25 | Splitless Precision Liner | Pack of 25 |
| 15548813 | Mixed | Mixed | 5 | Mixed Liner Pack | Pack of 5 |
| 16646912 | 4 mm | 78.5 mm | 5 | Double Taper Cyclo Liner | Pack of 5 |
| 30130381 | 2 mm | 120 mm | 1 | PTV Liner, Tenax TA | Pack of 5 |
Use promo code 28025
Log in to see stock.
BEST PRICE on Promo! Use promo code "28025" to get your promotional price.
Beschreibung
Use the gold-standard Thermo Scientific™ LinerGOLD™ GC Liners for consistently high accuracy, sensitivity, and precision in your gas chromatography (GC) analysis.
Use the gold standard in GC liner performance and consistency. LinerGOLD GC Liners provide enhanced inertness, leading to increased accuracy, sensitivity, and precision in your gas chromatography (GC) analysis. These injection port liners deliver:
- Increased accuracy and sensitivity in analysis, preventing the loss of sensitive compounds in analysis
- Lower detection limits when analyzing active and sensitive compounds
- Excellent reproducibility gives confidence in your results from liner to liner
- Consistently high level of performance time after time
LinerGOLD GC liners are manufactured and tested to ensure that you have confidence with every liner that is installed onto your GC system.
Spezifikation
Spezifikation
| Product Type | PTV Baffle Liner (Siltek) |
| Quantity | 5 |
| Length (Metric) | 120 mm |
| Packing Material | None |
| Diameter (Metric) Inner | 2 mm |
| Diameter (Metric) Outer | 2.75 mm |
| For Use With (Equipment) | Thermo Scientific Instruments |