Learn More
WTW™ Initial Calibration Kit
The QSC kit consists of three precision DIN buffers (pH 4.01, pH 6.87 and pH 9.18 with a deviation of +/- 0.01 pH at 25° C respectively) an initial calibration can be performed in glass vials for IDS pH electrodes. Ideal for quality control: All subsequent calibrations are compared to this calibration and thus deliver exactly the current status of the sensor with respect to its initial values.
Brand:  WTW™ 109830
Additional Details : Weight : 0.11970kg
Product Code. 11745174
  • 382.00 NOK / Each
Estimated Shipment: 17-06-2024
Specifications
Initial Calibration Kit
With pack of 3 disposable 20mL ampoules, pH 4.01, pH 6.86, pH 9.18
IDS pH electrodes; Includes; Pack of 3 disposable 20mL ampoules, pH 4.01, pH 6.86, pH 9.18