missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ING2 (Human) Recombinant Protein
Human ING2 full-length ORF ( AAH30128, 1 a.a. - 280 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003622-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MLGQQQQQLYSSAALLTGERSRLLTCYVQDYLECVESLPHDMQRNVSVLRELDNKYQETLKEIDDVYEKYKKEDDLNQKKRLQQLLQRALINSQELGDEKIQIVTQMLELVENRARQMELHSQCFQDPAESERASDKAKMDSSQPERSSRRPRRQRTSESRDLCHMANGIEDCDDQPPKEKKSKSAKKKKRSKAKQEREASPVEFAIDPNEPTYCLCNQVSYGEMIGCDNEQCPIEWFHFSCVSLTYKPKGKWYCPKCRGDNEKTMGKSTEKTKKDRRSR
Specifications
AAH30128 | |
inhibitor of growth family, member 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ING2 | |
GST |
3622 | |
Wheat germ expression system | |
10 μg | |
ING1L, p33ING2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |