Learn More
Masterflex™ Inert in-line Check Valves
Product Code. 11769609
Click to view available options
Material:
EPDM, PEEK
Unit Size:
Each
Product Code. 11769609
Brand: Masterflex™ 0135500
Only null left
Add to basket
Add to basket
Designed for use with Cole-Parmer tubing. In-line check valves feature a smooth flow path that minimizes shear and turbulence.
  • Inert in-line Outlet Check Valve
  • No metal components allow inert flow path
  • No back flow leakage with back pressure of 0 to 100psi
  • Ideal for high-purity, low-pressure applications
  • Zero maintenance
Specifications
Material EPDM, PEEK
Inlet 1/4 in. to 28 UNF
Outlet Port 1/4 in. to 28 UNF
Maximum Pressure 100 psi
Product Type Inert in-line Outlet Check Valve