Learn More
Buerkle™ Industrial Measuring Jug with Closed Handle
Product Code. 18080915 Shop All Bürkle Products
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
18080915 Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18080915 Supplier Buerkle™ Supplier No. 72042000
  • ONLINE EXCLUSIVE PRICE
    118.00 NOK
    112.65 NOK / Pack of 1
    Save 5.35 NOK
    4% Off
Estimated Shipment: 07-01-2027
The measuring jug has two measuring scales, embossed transparent and with blue lettering. With ergonomic handle, available in closed design for special stability or with open handle for stacking to save space.
  • PP transparent
  • With embossed graduation printed in blue
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Capacity (Metric) 2000 mL
Height (Metric) 20.6 cm
Product Type Measuring Jug