missing translation for 'onlineSavingsMsg'
Learn More

Indotecan, MedChemExpress

Product Code. p-200096480
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493527

Brand: Medchem Express HY18351

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Indotecan (LMP-400) is a potent topoisomerase 1(Top1) inhibitor with IC50 values of 300, 1200, 560 nM for P388, HCT116, MCF-7 cell lines, respectively.

Indotecan (LMP-400) is a potent topoisomerase 1(Top1) inhibitor with IC50 values of 300, 1200, 560 nM for P388, HCT116, MCF-7 cell lines, respectively.

Specifications

Chemical Name or Material Indotecan
CAS 915303-09-2
Color Gray
Purity Grade Notes Research
Molecular Formula C26H26N2O7
Quantity 50 mg
Synonym LMP-400 NSC-724998
Solubility Information DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES O=C1C2=CC(OC)=C(OC)C=C2C3=C(C(C=C(OCO4)C4=C5)=C5C3=O)N1CCCN6CCOCC6
Molecular Weight (g/mol) 478.49
Formula Weight 478.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.