Learn More
Indotecan, MedChemExpress
Product Code. 16493527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16493527 50 mg 50mg
16467508 10 mg 10mg
16459987 5 mg 5mg
16447466 100 mg 100mg
4 options
Product Code. 16493527
Brand: Medchem Express HY18351
MedChemExpress Indotecan (LMP-400) is a potent topoisomerase 1(Top1) inhibitor with IC50 values of 300, 1200, 560 nM for P388, HCT116, MCF-7 cell lines, respectively.
Indotecan (LMP-400) is a potent topoisomerase 1(Top1) inhibitor with IC50 values of 300, 1200, 560 nM for P388, HCT116, MCF-7 cell lines, respectively.
Specifications
Chemical Name or Material Indotecan
CAS 915303-09-2
Color Gray
Purity Grade Notes Research
Molecular Formula C26H26N2O7
Quantity 50 mg
Synonym LMP-400 NSC-724998
Solubility Information DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES O=C1C2=CC(OC)=C(OC)C=C2C3=C(C(C=C(OCO4)C4=C5)=C5C3=O)N1CCCN6CCOCC6
Molecular Weight (g/mol) 478.49
Formula Weight 478.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation