missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-acetic Acid, Millipore™

Product Code. 10485311
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10485311 100 g 100g
10046580 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10485311 Supplier Millipore Supplier No. 1003530100
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-51-4

Specifications

Melting Point 167°C to 170°C
MDL Number MFCD00005636
Quantity 100 g
Synonym Indole-3-acetic acid, JAA, Heteroauxine, Indole-3-acetic acid
Molecular Weight (g/mol) 175.18
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.