Learn More
Indium(III) Trifluoromethanesulfonate, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Indium(III) Trifluoromethanesulfonate |
| CAS | 128008-30-0 |
| Molecular Formula | C3F9InO9S3 |
| Quantity | 50 g |
| Synonym | Indium Triflate, Indium Trifluoromethylsulfonate, Indium Tris(trifluoromethanesulfonate), Indium Tritriflate, Indium(3+) Triflate, Indium(3+) Trifluoromethanesulfonate, Indium(III) Triflate, Indium(III) Trifluoromethanesulfonate, Trifluoromethanesulfonic Acid Indium(3+) Salt, Tris(trifluoromethanesulfonato)indium |
| InChI Formula | InChI=1S/3CHF3O3S.In/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3 |
| SMILES | [In+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F |
| IUPAC Name | indium(3+);trifluoromethanesulfonate |
| Molecular Weight (g/mol) | 562.03 |
| Formula Weight | 561.76 |