missing translation for 'onlineSavingsMsg'
Learn More

Indisulam, MedChemExpress

Product Code. 16458467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458467 5 mg 5mg
16481947 50 mg 50mg
16463376 25 mg 25mg
16455798 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16458467 Supplier Medchem Express Supplier No. HY13650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Indisulam (E 7070) is a carbonic anhydrase inhibitor with anticancer activity. Indisulam (E 7070) is a sulfonamide agent that targets the G1 phase of the cell cycle. Indisulam (E 7070) causes a blockade in the G1/S transition through inhibition of the activation of both CDK2 and cyclin E. Indisulam (E 7070) targets splicing by inducing RBM39 degradation via recruitment to DCAF15.

Indisulam (E 7070) is a carbonic anhydrase inhibitor with anticancer activity. Indisulam (E 7070) is a sulfonamide agent that targets the G1 phase of the cell cycle. Indisulam (E 7070) causes a blockade in the G1/S transition through inhibition of the activation of both CDK2 and cyclin E. Indisulam (E 7070) targets splicing by inducing RBM39 degradation via recruitment to DCAF15.

Specifications

Chemical Name or Material Indisulam
CAS 165668-41-7
Color Light Nude
Purity Grade Notes Research
Molecular Formula C14H12ClN3O4S2
Quantity 5 mg
Synonym E 7070
Solubility Information DMSO : 100 mg/mL (259.17 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(S(=O)(N)=O)C=C1)(NC2=CC=CC3=C2NC=C3Cl)=O
Molecular Weight (g/mol) 385.85
Formula Weight 385.85
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.