Learn More
Indane, 95%
Product Code. p-7025776
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
Product Code. Quantity unitSize
11449427 100 mL 100mL
11459427 500 mL 500mL
11469427 2500 mL 2500mL
3 options
Product Code. 11449427
Brand: Thermo Scientific Alfa Aesar B21442.AE
Request Bulk or Custom Format
CAS: 496-11-7 | C9H10 | 118.179 g/mol
Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.
Solubility
Soluble in ethanol, diethyl ether, chloroform (slightly), and ether. Insoluble in water.
Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 496-11-7
Molecular Formula C9H10
Molecular Weight (g/mol) 118.179
MDL Number MFCD00003795
InChI Key PQNFLJBBNBOBRQ-UHFFFAOYSA-N
Synonym indan, indane, hydrindene, benzocyclopentane, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, 1,2-hydrindene, hydrindonaphthene, indane alkane, indene, 2,3-dihydro
PubChem CID 10326
ChEBI CHEBI:37911
IUPAC Name 2,3-dihydro-1H-indene
SMILES C1CC2=CC=CC=C2C1
Specifications
Melting Point -51°C
Density 0.962
Boiling Point 175°C to 177°C
Flash Point 50°C (122°F)
Refractive Index 1.537
Quantity 100 mL
UN Number UN3295
Beilstein 1904376
Merck Index 14,4931
Solubility Information Soluble in ethanol,diethyl ether,chloroform (slightly),and ether. Insoluble in water.
Formula Weight 118.18
Percent Purity 95%
Chemical Name or Material Indane
Show More Show Less
compliance-icons
Product Identifier
  • Indane
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Indan