missing translation for 'onlineSavingsMsg'
Learn More

INAVA Antibody (OTI6A6), HRP, Novus Biologicals™

Product Code. 18101373 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18101373 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18101373 Supplier Novus Biologicals Supplier No. NBP272021H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

INAVA Monoclonal specifically detects INAVA in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INAVA
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6A6
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias C1orf106, chromosome 1 open reading frame 106, hypothetical protein LOC55765
Gene Symbols C1ORF106
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-270 of human C1orf106 (NP_060735) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55765.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.