Description
- Fits EX models 124-523
- AX 124, 124/E
- AX 223, 223/E, 223N/E
- AX 224, 224/E, 224N
- AX 324
- AX 423, 423/E, 423N, 423N/E
- AX 523/E, 523N/E
- Protects your scale from dust, fingerprints, and spills
Specifications
Specifications
| For Use With (Equipment) | Electronic Balance, AX Model |
| Languages | Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) |
| Product Line | In-Use Covers |
| Type | In-use Cover |