Learn More
Ohaus™ Impact Printer
Product Code. 13445129 Shop All Ohaus Products
Change view
Click to view available options
Dimensions (L x W x H):
180 x 106 x 88 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
13445129 180 x 106 x 88 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13445129 Supplier Ohaus™ Supplier No. 30064202
Only null left
Add to basket
Add to basket
Combines high performance and multiple functions to offer OHAUS quality and durability in a portable printer. Ohaus™ Impact Printers offer advanced functionality in an easy-to-use and set up printer at an exceptional value.
Ideal for use in conjunction with OHAUS balances and scales in laboratory, industrial, and educational applications to ensure accurate documentation of weighing results.
  • AC adapter included
  • RS233 communication
  • ABS Housing
  • Status indicator, 40 characters per line, compatible with continuous label paper, perform zero and tare operations directly from the printer
TRUSTED_SUSTAINABILITY
Specifications
Dimensions (L x W x H) 180 x 106 x 88 mm
Width (Metric) 1.6 mm
Height (Metric) 88 mm
Quantity 1 Each
Type Impact Printer