missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ImmunoCult™ Mouse Th2 Differentiation Supplement

Product Code. 17177641
Encompass
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 17177641

Brand: STEMCELL Technologies 10955

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Supplement for the differentiation of mouse naïve CD4+ T cells into Th2 cells

  • ImmunoCult™ Mouse Th2 Differentiation Supplement contains a combination of recombinant mouse cytokines and a blocking monoclonal antibody optimized to promote the differentiation of mouse naïve CD4+ T cells into Th2 cells
  • This supplement is intended for use with RPMI 1640 Medium (Catalog #36750) containing fetal bovine serum and other additives, as well as anti-mouse CD3 and anti-mouse CD28 monoclonal antibodies as activating agents
  • Research into the regulation of mouse naïve T cell differentiation and Th2 function
  • Development of procedures to expand and polarize naïve T cells toward a Th2 subset

Specifications

Cell Type T Cells, T Cells, CD4+
For Use With (Application) Cell Culture, Differentiation, Expansion
Quantity 1 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.