Learn More
STEMCELL Technologies ImmunoCult™ Human CD3/CD28 T Cell Activator
Product Code. 15553062
Encompass
Change view
Click to view available options
Quantity:
2 mL
5 x 2 mL
Unit Size:
2mL
Pack of 5
Product Code. Quantity unitSize
15553062 2 mL 2mL
16425234 5 x 2 mL Pack of 5
2 options
Product Code. 15553062
Brand: STEMCELL Technologies 10971
Human T cell activation and expansion reagent
  • Achieve robust activation and expansion of T cells in the absence of magnetic beads, feeder cells, or antigens
  • This product's gentle activation stimulus ensures a high viability of activated T cells, which can be further expanded, for example in ImmunoCult™-XF T Cell Expansion Medium (Catalog #10981)
  • Tetrameric antibody complexes bind to and cross-link CD3 and CD28 cell surface ligands, thereby providing the required signals for T cell activation
  • ImmunoCult™ Human CD3/CD28 T Cell Activator can be used on the Seahorse XF Analyzer to measure T cell activation response and is also available as part of the Agilent Seahorse XF Hu T Cell Activation Assay Kit
  • This product is designed for cell therapy research applications, but may be qualified for use as an ancillary material (AM) following the framework outlined in USP<1043>
  • STEMCELL Technologies can work with you to qualify this reagent as an AM under an approved investigational new drug (IND) or clinical trial application (CTA)
Specifications
Cell Type T Cells, T Cells, CD4+, T Cells, CD8+
For Use With (Application) Activation, Cell Culture, Expansion
Quantity 2 mL