Learn More
STEMCELL Technologies ImmunoCult™ Dendritic Cell Culture Kit
Description
- ImmunoCult™-ACF Dendritic Cell Medium and Supplements, sold individually or as a kit, have been developed for the in vitro culture and differentiation of human monocytes into dendritic cells (DCs)
- ImmunoCult™-ACF Dendritic Cell Medium is a serum-free and animal component-free medium optimized for the in vitro culture and differentiation of human monocytes into DCs
- ImmunoCult™-ACF Dendritic Cell Differentiation Supplement contains a combination of animal component-free recombinant human cytokines formulated to support the differentiation of immature DCs from human monocytes
- The ImmunoCult™ Dendritic Cell Maturation Supplement has been formulated to support maturation of immature DCs to mature DCs
Specifications
Specifications
| Quantity | 1 Kit |
| For Use With (Application) | Cell Culture, Differentiation |
| Cell Type | Dendritic Cells; Monocytes |