missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies ImmunoCult™-ACF Human B Cell Expansion Supplement
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
Description
- ImmunoCult™-ACF Human B Cell Expansion Supplement is a serum-free and animal component-free supplement formulated to promote robust in vitro expansion of human peripheral blood B cells in the absence of serum, feeder cells, or specialized culture plates
- The supplement contains recombinant proteins and chemically defined components that support the activation and expansion of human B cells and their maturation to plasma cells
- ImmunoCult™-ACF Human B Cell Expansion Supplement is a 50X supplement that can be added to a base medium such as ImmunoCult™-XF T Cell Expansion Medium (Catalog #10981) to prepare a complete serum-free medium for culturing and expanding human B cells
Specifications
Specifications
| Cell Type | B Cells |
| For Use With (Application) | Activation, Cell Culture, Differentiation, Expansion, Genome Editing |
| Quantity | 2 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction