Learn More
Thermo Scientific™ Immuno Standard Modules White and Black
BIOPHARMA
Product Code. 10060591
Change view
Click to view available options
Color:
Black
Clear
White
Volume (Metric) Well:
350 μL
380 μL
400 μL
Surface Treatment:
Hydrophobic
MaxiSorp
PolySorp
Unit Size:
Pack of 320
Pack of 60
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) Well Surface Treatment Unit Size
10060591 Black 400 μL PolySorp Pack of 60
10376231 White 350 μL MaxiSorp Pack of 60
10040591 White 350 μL PolySorp Pack of 60
10080611 Clear 350 μL MaxiSorp Pack of 60
10759081 Black 400 μL MaxiSorp Pack of 60
10634247 White 380 μL Hydrophobic Pack of 320
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10060591 Supplier Thermo Scientific™ Supplier No. 475523
  • ONLINE EXCLUSIVE PRICE
    4210.00 NOK
    4106.28 NOK / Pack of 60
    Save 103.72 NOK
    2% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Bulk Quote Request
Conduct fluorescence- and luminescence-based immunoassays and binding assays using these immuno standard modules white and black.
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.
Features:
  • Available in 8-, 12- or 16-well strips
  • Framed or unframed for choice of optimal assay
  • Flat-bottom or C-shaped wells
  • Choice of three surface types for optimal binding
  • White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
  • Black format to reduce backscatter light, which may be encountered in epifluorescence
  • Transparent or clear format for use in time-resolved fluorescence applications only
Choice of Surface:
  • Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
  • Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.
TRUSTED_SUSTAINABILITY
Specifications
Breakapart No
Binding Property Hydrophobic
Certified No
Format Standard
Detection Method Fluorescence
Array 8 x 12
Packaging 60 Cases
Binding Type Adsorption
Color Black
Product Type Microplate Strip
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 350 μL
Sterility Non-sterile
Surface Treatment PolySorp
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Fluorescence
No. per Case 60
No. per Pack 5
Plate Blocking None
Quantity 60/Cs.
Target Molecule Lipids
Show More Show Less
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?
Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.