Learn More
Hughes Immersion Heated 350 L Tank Shower
Product Code. 17792348
Change view
Click to view available options
Capacity (Metric):
350 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17792348 350 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17792348 Supplier Hughes Supplier No. HSHM14K3SS
  • ONLINE EXCLUSIVE PRICE
    124036.29 NOK
    77275.43 NOK / Pack of 1
    Save 46760.86 NOK
    37% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Immersion heated 350 liter emergency tank shower with galvanized mild steel framework.
  • Immersion heated emergency tank shower for use in nonhazardous environments
  • Suitable for use where insufficient water pressure exists
  • An integral immersion heater, controlled by a dual safety thermostat, maintains the water in the tank at a constant temperature
  • Side mounted eye/face wash unit
Specifications
Capacity (Metric) 350 L
Material Stainless Steel
Product Type Tank Shower