missing translation for 'onlineSavingsMsg'
Learn More

Imiquimod, MedChemExpress

Product Code. p-200096350
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16428156

Brand: Medchem Express HYB0180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Imiquimod (R 837), an immune response modifier, is a selective toll like receptor 7 (TLR7) agonist. Imiquimod exhibits antiviral and antitumor effects in vivo. Imiquimod can be used for the research of external genital, perianal warts, cancer and COVID-19.

Imiquimod (R 837), an immune response modifier, is a selective toll like receptor 7 (TLR7) agonist. Imiquimod exhibits antiviral and antitumor effects in vivo. Imiquimod can be used for the research of external genital, perianal warts, cancer and COVID-19.

Specifications

Chemical Name or Material Imiquimod
Physical Form Powder
CAS 99011-02-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H16N4
For Use With (Application) COVID-19-anti-virus
Synonym R 837
Solubility Information H2O : 2.64 mg/mL (10.99 mM; ultrasonic and adjust pH to 2 with HCl) ∣DMSO : 2 mg/mL (8.32 mM; ultrasonic and warming and heat to 70°C)
SMILES NC1=NC2=CC=CC=C2C3=C1N=CN3CC(C)C
Molecular Weight (g/mol) 240.3
Formula Weight 240.3
Percent Purity 98.08%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.