Learn More
Imiquimod hydrochloride, MedChemExpress
Product Code. 16438156
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Product Code. Quantity unitSize
16438156 100 mg 100mg
16411396 200 mg 200mg
16402056 500 mg 500mg
3 options
Product Code. 16438156
Brand: Medchem Express HYB0180A
MedChemExpress Imiquimod hydrochloride (R 837 hydrochloride), an immune response modifier, is a selective toll like receptor 7 (TLR7) agonist. Imiquimod hydrochloride exhibits antiviral and antitumor effects in vivo. Imiquimod hydrochloride can be used for the research of external genital, perianal warts, cancer and COVID-19.
Imiquimod hydrochloride (R 837 hydrochloride), an immune response modifier, is a selective toll like receptor 7 (TLR7) agonist. Imiquimod hydrochloride exhibits antiviral and antitumor effects in vivo. Imiquimod hydrochloride can be used for the research of external genital, perianal warts, cancer and COVID-19.
Specifications
Chemical Name or Material Imiquimod hydrochloride
Physical Form Solid
CAS 99011-78-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H17ClN4
For Use With (Application) COVID-19-anti-virus
Synonym R 837 hydrochloride
Solubility Information DMSO : 8 mg/mL (28.91 mM; ultrasonic and warming and heat to 50°C) ∣H2O : 4.8 mg/mL (17.34 mM; Need ultrasonic) ∣Ethanol : 3.85 mg/mL (13.91 mM; Need ultrasonic)
SMILES NC1=NC2=CC=CC=C2C3=C1N=CN3CC(C)C.Cl
Molecular Weight (g/mol) 276.76
Formula Weight 276.76
Percent Purity 99.76%
Grade Research