Learn More
Imidazole, TRC
Product Code. 30011631
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011631 25 g 25g
30011475 1 g 1g
30003326 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011631 Supplier Toronto Research Chemicals Supplier No. TRCI35020025G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 288-32-4
Molecular Formula C3 H4 N2
Molecular Weight (g/mol) 68.08
Synonym 1H-Imidazole, Imidazole, Bifonazole Imp. C (EP), Ondansetron Hydrochloride Dihydrate Imp. E (EP), Clotrimazole Imp. D (EP), Enalapril Maleate Imp. I (EP), Zoledronic Acid Monohydrate Imp. C (Pharmeuropa), 1H-Imidazole (9CI, ACI), Imidazole (8CI), 1,3-Diaza-2,4-cyclopentadiene, 1,3-Diazole, 3-Azapyrrole, Curezol SIZ, Glyoxalin, Glyoxaline, Imidazol, Imutex, Methanimidamide, N,N'-1,2-ethenediyl-, Miazole, NSC 60522
IUPAC Name 1H-imidazole
SMILES c1c[nH]cn1
Specifications
Quantity 25 g
InChI Formula InChI=1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)
Formula Weight 68.0374
Percent Purity >95%
Chemical Name or Material Imidazole
Grade HPLC