Learn More
Imidazol-1-yl-acetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Imidazol-1-yl-acetic Acid |
| CAS | 22884-10-2 |
| Molecular Formula | C5 H6 N2 O2 |
| Quantity | 5 g |
| Synonym | 2-(1H-Imidazol-1-yl)acetic Acid, Imidazole-1-acetic acid (6CI,7CI,8CI), (1H-Imidazol-1-yl)acetic acid, 2-(1H-Imidazol-1-yl)acetic acid, Imidazol-1-ylacetic acid |
| InChI Formula | InChI=1S/C5H6N2O2/c8-5(9)3-7-2-1-6-4-7/h1-2,4H,3H2,(H,8,9) |
| SMILES | OC(=O)Cn1ccnc1 |
| IUPAC Name | 2-imidazol-1-ylacetic acid |
| Molecular Weight (g/mol) | 126.11 |
| Formula Weight | 126.0429 |