Learn More
Imidazo[1,2-a]pyridine-6-carboxylic acid, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 139022-25-6 |
| Molecular Formula | C8H6N2O2 |
| MDL Number | MFCD07021498 |
| Quantity | 5 g |
| SMILES | O=C(C1=CN2C(C=C1)=NC=C2)O |
| Molecular Weight (g/mol) | 162.15 |
| Percent Purity | 97% |
| Chemical Name or Material | Imidazo[1,2-a]pyridine-6-carboxylic acid |