Description
- Manufactured from borosilicate glass.
- Complies with DIN 12672 standards.
- With or without stopcock
- Graduated to 100mL or 1000mL
- Ring mark at 1000mL
- *1000mL graduated version does not include ring mark.
Specifications
Specifications
| Material | Borosilicate Glass |
| Graduated | Yes |
| Graduations | to 100 mL, Ring Mark at 1000 mL |
| Stopcock | Yes |
| Product Type | Sedimentation cone |