missing translation for 'onlineSavingsMsg'
Learn More

IMD-0354, MedChemExpress

Product Code. p-200098775
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16463528

Brand: Medchem Express HY10172

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IMD-0354 (IKK2 Inhibitor V) is a selective IKKβ inhibitor which inhibits NF-κB activity. IMD0354 inhibits TNF-α induced NF-κB transcription activity with an IC50 of 1.2 uM.

IMD-0354 (IKK2 Inhibitor V) is a selective IKKβ inhibitor which inhibits NF-κB activity. IMD0354 inhibits TNF-α induced NF-κB transcription activity with an IC50 of 1.2 uM.

Specifications

Chemical Name or Material IMD-0354
CAS 978-62-1
Color White
Purity Grade Notes Research
Molecular Formula C15H8ClF6NO2
Quantity 10 mg
Synonym IKK2 Inhibitor V
Solubility Information DMSO : ≥ 100 mg/mL (260.64 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(NC1=CC(C(F)(F)F)=CC(C(F)(F)F)=C1)C2=CC(Cl)=CC=C2O
Molecular Weight (g/mol) 383.67
Formula Weight 383.67
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.