Learn More
Invitrogen™ Image-iT™ Plant Tissue Hard-Set Mountant
Product Code. 16319583
Click to view available options
Capacity:
30 mL
100 mL
Unit Size:
100mL
30mL
Product Code. 16319583
Brand: Invitrogen™ V11330
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Image-iT Plant Tissue Hard-Set Mountant is used for imaging, storage, and archiving of plant tissues optically cleared with Image-iT Plant Tissue Clearing Reagent.
These reagents are compatible with both fluorescence-based and colorimetric-based applications. Image-iT Plant Tissue mountant is based on Visikol™ Mount, which has a history of use in fluorescent and colorimetric studies with plants.
• Clears and mounts plant tissue for whole specimen imaging
• Hard-set mountant with refractive index of 1.415
TRUSTED_SUSTAINABILITY
Specifications
Capacity 100 mL
Content And Storage Store at 15°C to 25°C. Do not freeze.
When stored as directed, product is stable for (at least 6 months after receipt.
)
Product Line Image-iT
Shipping Condition Room Temperature