Learn More
Thermo Scientific™ Image-iT™ Cell Painting Kit
Shop All Thermo Scientific ProductsDescription
The Image-iT Cell Painting Kit is a collection of reagents that can be used to check cell health/cytotoxicity by “painting” different cellular components. When combined with a high-content imager, such as the CellInsight platform, these reagents can provide a phenotypical readout of cell status. This kit is based on the publication by Bray et al. (1) describing the morphological profiling of cells using multi-plexed fluorescent dyes.
This kit includes the following reagents (1 each):
• Hoechst 34580, Trihydrochloride, Trihydrate (Cat. No. H21486)
• Concanavalin A, Alexa Fluor 488 Conjugate (Cat. No. C11252)
• Wheat Germ Agglutinin, Alexa Fluor 555 Conjugate (Cat. No. W32464)
• Alexa Fluor 568 Phalloidin (Cat. No. A12380)
• SYTO 14 Green Fluorescent Nucleic Acid Stain (Cat. No. S7576)
• MitoTracker Deep Red FM (Cat. No. M22426)
Reference
1. Bray MA, Singh S, Han H, Davis CT, Borgeson B, Hartland C, Kost-Alimova M, Gustafsdottir SM, Gibson CC, Carpenter AE. Cell Painting, a high-content image-based assay for morphological profiling using multiplexed fluorescent dyes. Nat Protoc. 2016 Sep;11(9):1757-74.
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| High-throughput Compatibility | Yes |
| Quantity | 1 Kit |