missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ILVBL Polyclonal specifically detects ILVBL in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ILVBL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Dilution | Western Blot 1:1000 |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Accession No. | A1L0T0 |
| Gene Alias | acetolactate synthase homolog, acetolactate synthase-like protein, AHASFLJ39061,209L8, EC 2.2.1, EC 2.2.1.-, ILV2H, ilvB (bacterial acetolactate synthase)-like, IlvB-like protein, MGC1269, MGC19535 |
| Gene Symbols | ILVBL |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?