Learn More
Ilorasertib, MedChemExpress
Product Code. 16433027
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Product Code. 16433027
Brand: Medchem Express HY16018
MedChemExpress Ilorasertib (ABT-348) is a potent and ATP-competitive multitargeted kinase inhibitor, which inhibits Aurora C, Aurora B, and Aurora A with IC50s of 1 nM, 7 nM, 120 nM, respectively. Ilorasertib also suppresses RET tyrosine kinase, PDGFRβ and Flt1 with IC50s of 7 nM, 3 nM and 32 nM, respectively.
Ilorasertib (ABT-348) is a potent and ATP-competitive multitargeted kinase inhibitor, which inhibits Aurora C, Aurora B, and Aurora A with IC50s of 1 nM, 7 nM, 120 nM, respectively. Ilorasertib also suppresses RET tyrosine kinase, PDGFRβ and Flt1 with IC50s of 7 nM, 3 nM and 32 nM, respectively.
Specifications
Chemical Name or Material Ilorasertib
CAS 1227939-82-3
Color Brown
Purity Grade Notes Research
Molecular Formula C25H21FN6O2S
Quantity 50 mg
Synonym ABT-348
Solubility Information DMSO : 41.67 mg/mL (85.29 mM; Need ultrasonic)
SMILES O=C(NC1=CC=CC(F)=C1)NC2=CC=C(C3=CSC4=C3C(N)=NC=C4C5=CN(CCO)N=C5)C=C2
Molecular Weight (g/mol) 488.54
Formula Weight 488.54
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.