Learn More
Cytiva TempliPhi™ Large Construct Kit
Product Code. 10362710 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
1000
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10362710 1000 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10362710 Supplier Cytiva Supplier No. 25640080
Includes
Sample buffer, Reaction buffer, and Enzyme mix
Uses isothermal rolling circle amplification (RCA) to efficiently prepare DNA specifically from large constructs (e.g., fosmids and BACs) sufficient for DNA sequencing
  • Start from microliter quantities of purified templates, as well as bacterial culture or glycerol stocks
  • Generate 5μg amounts of template DNA for use directly in cycle sequencing reactions without purification
  • Simplified process, 20 minutes hands-on time
  • Amplification is completed in 18 hours with no need for thermal cycling
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.
Description illustra TempliPhi Large Construct Kit uses isothermal rolling circle amplification (RCA) to efficiently prepare DNA sequencing templates specifically from large DNA vectors (e.g., fosmids and BACs).
For Use With (Application) Sequencing Template Amplification Kits and Reagents
Includes 9mL Sample Buffer, 9mL Reaction Buffer, 500μL Enzyme mix
No. of Reactions 1000
Product Type illustra TempliPhi Large Construct Kit
Test Time 18 h
compliance-icons
Product Identifier
  • TempliPhi
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P405-Store locked up.